Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dup
Entrez Gene ID
47121
Gene Name
double parked
Gene Aliases
19k, CDT1, CDT1/DUP, CG8171, Cdt1, DUP, DUP/CDT1, Dmel\CG8171, Dmel_CG8171, Dup, anon-WO0257455.31, cdt1, cdt1/dup, fs(2)51Fa, fs(2)PA77, fs(2)ltoPA77, l(2)51Ec, l(2)k03308
Chromosome Location
Chr.2R: 15376619 - 15380988 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000996
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299522.1 NP_001286451.1 1 - 2 297 72
NM_080139.5 NP_524878.1 1 - 2 671 72
GenBank mRNA AF279146.1 - 1 - 2 293 72
AY118555.1 - 1 - 2 633 72

View Full Product Details