Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ebo
Entrez Gene ID
47141
Gene Name
ellipsoid body open
Gene Aliases
CG3923, Dmel\CG3923, Dmel_CG3923, EBO, EG:165H7.3, Exp1, Exp6
Chromosome Location
Chr.X: 471019 - 476579 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266572
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297809.1 NP_001284738.1 3 - 4 866 69
NM_001297810.1 NP_001284739.1 3 - 4 866 69
NM_080140.3 NP_524879.2 3 - 4 866 69
GenBank mRNA BT132921.1 - 3 - 4 834 69

View Full Product Details