Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fkbp59
Entrez Gene ID
47762
Gene Name
FK506-binding protein 59kD
Gene Aliases
CG4535, Dmel\CG4535, Dmel_CG4535, FKBP59, FKBP59-bp1, anon-WO0118547.161, dFKBP4, dFKBP59, fkbp59, l(2)k00424, l(2)k09010
Chromosome Location
Chr.2L: 9888005 - 9890119 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029174
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298855.1 NP_001285784.1 3 - 4 600 51
NM_080156.4 NP_524895.2 3 - 4 507 51
GenBank mRNA AY058729.1 - 3 - 4 496 51

View Full Product Details