Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mei-9
Entrez Gene ID
31373
Gene Name
meiotic 9
Gene Aliases
CG3697, DhR1, DmXPF, Dmel\CG3697, Dmel_CG3697, MEI-9, Mei-9, Rad1p, mei9, mus(1)110, mus110, xpf/mei-9
Chromosome Location
Chr.X: 4314230 - 4318801 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002707
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297930.1 NP_001284859.1 6 - 7 2996 82
NM_080329.3 NP_525068.1 7 - 8 3111 82
GenBank mRNA BT150257.1 - 7 - 8 3111 82

View Full Product Details