Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
drm
Entrez Gene ID
49638
Gene Name
drumstick
Gene Aliases
CG10016, Dmel\CG10016, Dmel_CG10016, drum
Chromosome Location
Chr.2L: 3539247 - 3548423 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024244
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080527.3 NP_536788.2 2 - 3 877 146
NM_164545.2 NP_722921.1 2 - 3 977 146
GenBank mRNA AY070883.1 - 2 - 3 923 146

View Full Product Details