Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Him
Entrez Gene ID
32801
Gene Name
Holes in muscle
Gene Aliases
CG15064, Dmel\CG15064, Dmel_CG15064, him
Chromosome Location
Chr.X: 18209427 - 18210741 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030900
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133045.4 NP_573273.1 1 - 2 394 100
GenBank mRNA AY070599.1 - 1 - 2 390 100

View Full Product Details