Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- wrapper
- Entrez Gene ID
-
37555
- Gene Name
- CG10382-PA
- Gene Aliases
-
CG10382, CT19059, Dmel\CG10382, Dmel_CG10382, Wr, Wrapper
- Chromosome Location
-
Chr.2R: 22379637 - 22382675 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0025878
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058056.4 |
NP_477404.1 |
1 - 2 |
348 |
89 |
GenBank mRNA |
AY047524.1 |
- |
1 - 2 |
473 |
89 |
View Full Product Details