Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or22b
- Entrez Gene ID
-
33336
- Gene Name
- Odorant receptor 22b
- Gene Aliases
-
22A.2, 22b, AN12, CG4231, DOR22A.2, DOR22b, DOR67, Dmel\CG4231, Dmel_CG4231, Dor67, OR22b, Or22, Or22A.2, Or22a/b, Or67, dOr22b, dor67, or22b
- Chromosome Location
-
Chr.2L: 1522697 - 1524257 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026397
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058077.3 |
NP_477425.1 |
3 - 4 |
1136 |
148 |
View Full Product Details