Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Galphai
- Entrez Gene ID
-
38765
- Gene Name
- G protein alpha i subunit
- Gene Aliases
-
CG10060, DG[[alpha1]], DGalpha1, Dmel\CG10060, Dmel_CG10060, G-alpha-65A, G-ia65A, G-ialpha65A, G-ialpha65a, G-oalpha65A, GNAI1/2/3, G[[alphai]], G[[i]], G[[i]]alpha, Gai, Galpha i, Galpha65A, Galpha[[i65A]], Galpha[[i]], Galpha[[i]]65A, Galphai/o, Galphaiota65A, Gi, Gi-alpha, Gialpha, galphai, i
- Chromosome Location
-
Chr.3L: 6972750 - 6980044 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001104
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058154.5 |
NP_477502.1 |
3 - 4 |
1091 |
63 |
GenBank mRNA |
AY051670.1 |
- |
3 - 4 |
1072 |
63 |
View Full Product Details