Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- sns
- Entrez Gene ID
-
44097
- Gene Name
- sticks and stones
- Gene Aliases
-
43-49, CG12495, CG13752, CG13753, CG13754, CG13755, CG18464, CG2385, CG33141, CG8278, CT33230, CT33232, Dmel\CG33141, Dmel_CG33141, SNS, Sns, rost, sns20
- Chromosome Location
-
Chr.2R: 8798489 - 8856091 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024189
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176106.2 |
NP_788286.1 |
16 - 17 |
3748 |
66 |
View Full Product Details