Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mthl6
- Entrez Gene ID
-
38839
- Gene Name
- methuselah-like 6
- Gene Aliases
-
CG16992, Dmel\CG16992, Dmel_CG16992, anon-WO0170980.127, anon-WO0170980.128, mth16
- Chromosome Location
-
Chr.3L: 7485967 - 7487918 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035789
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176295.2 |
NP_788473.1 |
5 - 6 |
1300 |
130 |
View Full Product Details