Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mthl6
Entrez Gene ID
38839
Gene Name
methuselah-like 6
Gene Aliases
CG16992, Dmel\CG16992, Dmel_CG16992, anon-WO0170980.127, anon-WO0170980.128, mth16
Chromosome Location
Chr.3L: 7485967 - 7487918 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035789
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176295.2 NP_788473.1 6 - 7 1506 98

View Full Product Details