Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nca
Entrez Gene ID
40186
Gene Name
Neurocalcin
Gene Aliases
CG7641, Dmel\CG7641, Dmel_CG7641, Ncalc, nca
Chromosome Location
Chr.3L: 19991887 - 19997568 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013303
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176365.3 NP_788543.1 3 - 4 315 79
GenBank mRNA AY071612.1 - 3 - 4 279 79
U15735.1 - 1 - 2 71 79

View Full Product Details