Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cm
Entrez Gene ID
31647
Gene Name
carmine
Gene Aliases
AP-3, AP-3mu, AP3mu3, CG3035, CG32731, Dmel\CG3035, Dmel_CG3035, Dmu3, Mu3, ap-3, apm3, cg3035, l(1)VEM119, mu3
Chromosome Location
Chr.X: 7009006 - 7011145 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000330
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272373.1 NP_001259302.1 3 - 4 501 62
NM_176700.3 NP_788873.1 3 - 4 501 62
GenBank mRNA AF110233.1 - 3 - 4 487 62
AJ009657.1 - 3 - 4 476 62
AY069388.1 - 3 - 4 477 62

View Full Product Details