Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- janB
- Entrez Gene ID
-
43568
- Gene Name
- janus B
- Gene Aliases
-
CG31024, CG7931, Dmel\CG7931, Dmel_CG7931, Jan, Jan-B, bs30f02.y1, jan, jan B, janus
- Chromosome Location
-
Chr.3R: 30037984 - 30038728 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001281
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057236.5 |
NP_476584.2 |
2 - 3 |
302 |
59 |
GenBank mRNA |
BT125584.1 |
- |
2 - 3 |
302 |
59 |
View Full Product Details