Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- nau
- Entrez Gene ID
-
42799
- Gene Name
- nautilus
- Gene Aliases
-
CG10250, Dmel\CG10250, Dmel_CG10250, Dmyd, MYOD, Mdrf, MyoD, NAU, Nau
- Chromosome Location
-
Chr.3R: 23712814 - 23718357 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002922
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057302.3 |
NP_476650.1 |
3 - 4 |
1123 |
152 |
View Full Product Details