Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slou
Entrez Gene ID
42547
Gene Name
slouch
Gene Aliases
CG6534, Dmel\CG6534, Dmel_CG6534, NK-1, NK1, NK1/S59, Nk1, S-59, S59, S59/NK-1, Slou, prd9, s59, slou/NK1
Chromosome Location
Chr.3R: 21548661 - 21557182 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002941
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275879.1 NP_001262808.1 3 - 4 2091 65
NM_057309.3 NP_476657.1 3 - 4 2091 65
GenBank mRNA X55393.1 - - 2092 65

View Full Product Details