Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- spn-B
- Entrez Gene ID
-
41746
- Gene Name
- spindle B
- Gene Aliases
-
CG3325, Dmel\CG3325, Dmel_CG3325, Rad51, Rad51-like, Spn-B, SpnB, XRCC3, rad51-like, spnB
- Chromosome Location
-
Chr.3R: 14328051 - 14329608 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003480
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057392.5 |
NP_476740.1 |
1 - 2 |
212 |
79 |
GenBank mRNA |
BT044133.1 |
- |
1 - 2 |
199 |
79 |
View Full Product Details