Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ask1
Entrez Gene ID
42366
Gene Name
Apoptotic signal-regulating kinase 1
Gene Aliases
ASK1, CG4720, D-ASK, DASK, DASK1, DPk92B/ASK1, Dmel\CG4720, Dmel_CG4720, MEKK5/ASK1, Pk92B, dASK, dASK1, dAsk1, pk92B
Chromosome Location
Chr.3R: 19879719 - 19886030 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014006
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057741.4 NP_477089.3 1 - 2 319 71
NM_206518.2 NP_996240.1 1 - 2 319 71

View Full Product Details