Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sodh-1
Entrez Gene ID
40836
Gene Name
Sorbitol dehydrogenase 1
Gene Aliases
BG:DS00464.2, CG1982, DM_7298873, Dmel\CG1982, Dmel_CG1982, Sdh-1, Sodh
Chromosome Location
Chr.3R: 7052335 - 7054541 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024289
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300274.1 NP_001287203.1 2 - 3 156 69
NM_058000.3 NP_477348.1 2 - 3 348 69
GenBank mRNA BT044539.1 - 2 - 3 348 69

View Full Product Details