Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vha13
Entrez Gene ID
42341
Gene Name
Vacuolar H[+] ATPase 13kD subunit
Gene Aliases
ATP6V1G, BcDNA:AT14009, CG6213, Dmel\CG6213, Dmel_CG6213, VhaG, chr3R:15469897..15470039, cnj, conjoined, l(3)05113, vha13
Chromosome Location
Chr.3R: 19643018 - 19644386 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283536
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300478.1 NP_001287407.1 1 - 2 181 67
NM_058089.5 NP_477437.1 1 - 2 181 67
GenBank mRNA AF143200.1 - 1 - 2 156 67
AY075202.1 - 1 - 2 199 67

View Full Product Details