Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CSN5
- Entrez Gene ID
-
42000
- Gene Name
- COP9 signalosome subunit 5
- Gene Aliases
-
BcDNA.LD14392, BcDNA:LD14392, CG14884, CH5, CSN-5, Csn5, DCH5, Dch5, Dmel\CG14884, Dmel_CG14884, JAB1, JAB1/CSN5, Jab1, JadBp, Q9XZ58, csn5, dch5, jab1/csn5, l(3)89Cf, l(3)L4032, l(3)M203, quo
- Chromosome Location
-
Chr.3R: 16470099 - 16471406 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027053
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058094.5 |
NP_477442.1 |
2 - 3 |
915 |
60 |
View Full Product Details