Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CstF64
- Entrez Gene ID
-
42239
- Gene Name
- Cleavage stimulation factor 64 kD subunit
- Gene Aliases
-
CG7697, CStF 64, CstF 64, CstF-64, Dmel\CG7697, Dmel_CG7697, dCstF64
- Chromosome Location
-
Chr.3R: 18659618 - 18661126 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027841
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058105.5 |
NP_477453.1 |
1 - 2 |
220 |
74 |
GenBank mRNA |
BT044537.1 |
- |
1 - 2 |
198 |
74 |
View Full Product Details