Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mst85C
Entrez Gene ID
41089
Gene Name
CG11993-PA
Gene Aliases
CG11993, Dmel\CG11993, Dmel_CG11993
Chromosome Location
Chr.3R: 9048645 - 9052231 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028708
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275470.1 NP_001262399.1 2 - 3 281 81
NM_058128.5 NP_477476.1 1 - 2 303 81
GenBank mRNA AY071454.1 - 1 - 2 300 81

View Full Product Details