Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nep2
Entrez Gene ID
40588
Gene Name
Neprilysin 2
Gene Aliases
BcDNA:GH07643, CG9761, DmNEP2, Dmel\CG9761, Dmel_CG9761, NEP2, dNEP2, nep2
Chromosome Location
Chr.3R: 4719458 - 4731878 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027570
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259985.2 NP_001246914.1 4 - 5 1228 71
NM_001275331.1 NP_001262260.1 4 - 5 1228 71
NM_079503.4 NP_524227.1 4 - 5 1228 71
GenBank mRNA AF132153.1 - 4 - 5 1202 71

View Full Product Details