Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lap
Entrez Gene ID
40863
Gene Name
like-AP180
Gene Aliases
AP180, BcDNA:GM10787, CG2520, Dmel\CG2520, Dmel_CG2520, EP(3)3060, EP3060, LAP, Lap, dAP-3, l(3)neo34, lap/AP180, sig
Chromosome Location
Chr.3R: 7186994 - 7200941 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086372
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144547.3 NP_001138019.2 7 - 8 1226 81
NM_001170067.2 NP_001163538.1 7 - 8 1226 81
NM_001260029.2 NP_001246958.1 7 - 8 1226 81
NM_001260030.2 NP_001246959.1 7 - 8 1226 81
NM_001260031.2 NP_001246960.1 7 - 8 1226 81
NM_001275409.1 NP_001262338.1 6 - 7 1193 81
NM_001275410.1 NP_001262339.1 7 - 8 1226 81
NM_001275411.1 NP_001262340.1 6 - 7 1193 81
NM_079528.3 NP_524252.2 7 - 8 1226 81

View Full Product Details