Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- alpha-Est7
- Entrez Gene ID
-
40901
- Gene Name
- alpha-Esterase-7
- Gene Aliases
-
Ali, Alpha-Est7, CG1112, DmalphaE7, Dmel\CG1112, Dmel_CG1112, EST23, Est-23, Est23, H, Mce, aE7, ali, alpha est7, alphaE7, alphaEst7, alphaest-7
- Chromosome Location
-
Chr.3R: 7519563 - 7523753 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015575
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079537.4 |
NP_524261.1 |
3 - 4 |
619 |
89 |
GenBank mRNA |
AY051473.1 |
- |
3 - 4 |
599 |
89 |
View Full Product Details