Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or85f
- Entrez Gene ID
-
41119
- Gene Name
- Odorant receptor 85f
- Gene Aliases
-
85D.1, 85f, CG16755, DOR32, DmOr85f, Dmel\CG16755, Dmel_CG16755, OR85f
- Chromosome Location
-
Chr.3R: 9398190 - 9399551 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037685
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079565.3 |
NP_524289.1 |
1 - 2 |
766 |
79 |
View Full Product Details