Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mRpL11
- Entrez Gene ID
-
41757
- Gene Name
- mitochondrial ribosomal protein L11
- Gene Aliases
-
BcDNA:RE67638, CG3351, Dmel\CG3351, Dmel_CG3351, L11, rpl-11
- Chromosome Location
-
Chr.3R: 14511838 - 14512691 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038234
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079627.3 |
NP_524351.1 |
2 - 3 |
511 |
64 |
GenBank mRNA |
AY094892.1 |
- |
2 - 3 |
482 |
64 |
View Full Product Details