Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cog2
- Entrez Gene ID
-
41870
- Gene Name
- Component of oligomeric golgi complex 2
- Gene Aliases
-
CG6177, Dmel\CG6177, Dmel_CG6177, ldl-Cp, ldlCp
- Chromosome Location
-
Chr.3R: 15358481 - 15361657 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026634
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079642.3 |
NP_524366.1 |
3 - 4 |
1928 |
79 |
View Full Product Details