Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cher
Entrez Gene ID
42066
Gene Name
cheerio
Gene Aliases
CG3937, Cher, Dmel\CG3937, Dmel_CG3937, FLN, FLN90, Fil, filamin, joy, sko
Chromosome Location
Chr.3R: 17091533 - 17125389 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014141
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144595.2 NP_001138067.1 4 - 5 881 63
NM_001202309.1 NP_001189238.1 14 - 15 4779 63
NM_001275727.1 NP_001262656.1 3 - 4 560 63
NM_001275728.1 NP_001262657.2 16 - 17 5656 63
NM_001275729.1 NP_001262658.1 15 - 16 5346 63
NM_001275730.1 NP_001262659.2 17 - 18 5671 63
NM_079659.4 NP_524383.3 14 - 15 4840 63
NM_169746.3 NP_732207.1 3 - 4 498 63
NM_169747.2 NP_732208.2 4 - 5 781 63
NM_206502.3 NP_996224.2 15 - 16 5407 63
GenBank mRNA AF188360.1 - 14 - 15 4816 63
AF188361.1 - 3 - 4 467 63
BT133034.1 - 4 - 5 908 63

View Full Product Details