Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lgr1
Entrez Gene ID
42133
Gene Name
Leucine-rich repeat-containing G protein-coupled receptor 1
Gene Aliases
CG7665, DLGR-1, DLGR1, Dmel\CG7665, Dmel_CG7665, FSH, FSH-TSH, Fsh, Fsh/CG7665, LGR1, LH/CG receptor, RH44949p, TSH, dLGR1
Chromosome Location
Chr.3R: 17725777 - 17733207 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016650
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079669.3 NP_524393.2 15 - 16 2381 59
NM_169777.2 NP_732265.1 15 - 16 2451 59
GenBank mRNA BT128700.1 - 15 - 16 2436 59

View Full Product Details