Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
how
Entrez Gene ID
42596
Gene Name
held out wings
Gene Aliases
0904/17, 24B, CG10293, Dmel\CG10293, Dmel_CG10293, HOW, How, KH93F, P62, SZ1, Who/How, anon-EST:Liang-2.39, clone 2.39, l(3)S053606, l(3)S090417, l(3)j5B5, l(3)j5D5, l(3)s2612, qkr, qkr[93F], sci, stru, who
Chromosome Location
Chr.3R: 22042191 - 22079321 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264491
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032032.3 NP_001027203.1 5 - 6 1031 74
NM_001170212.2 NP_001163683.1 5 - 6 1031 74
NM_001170213.2 NP_001163684.1 5 - 6 1031 74
NM_001275893.1 NP_001262822.1 5 - 6 1031 74
NM_079723.4 NP_524447.2 5 - 6 1031 74
NM_169992.3 NP_732695.1 5 - 6 1031 74
GenBank mRNA BT033005.1 - 5 - 6 988 74
U72331.1 - 5 - 6 1027 74
U85651.1 - 5 - 6 1010 74

View Full Product Details