Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rtp
Entrez Gene ID
40627
Gene Name
retinophilin
Gene Aliases
9C4, CG10233, Dmel\CG10233, Dmel_CG10233, UTA, anon-83ABa, retin, uta
Chromosome Location
Chr.3R: 5236092 - 5237315 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0087005
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141263.3 NP_649520.1 1 - 2 85 72
GenBank mRNA AY060630.1 - 1 - 2 42 72
DQ324736.1 - 1 - 2 48 72

View Full Product Details