Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLDIP2
Entrez Gene ID
40655
Gene Name
Polymerase (DNA-directed), delta interacting protein 2
Gene Aliases
CG12162, Dmel\CG12162, Dmel_CG12162
Chromosome Location
Chr.3R: 5467778 - 5469566 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037329
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141283.4 NP_649540.1 2 - 3 801 69
NM_169054.2 NP_730934.1 1 - 2 818 69
GenBank mRNA AY069102.1 - 2 - 3 749 69

View Full Product Details