Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hcs
Entrez Gene ID
40659
Gene Name
Holocarboxylase synthetase
Gene Aliases
CG14670, Dmel\CG14670, Dmel_CG14670, HCS, HSC, NEST:bs27h05
Chromosome Location
Chr.3R: 5474899 - 5485393 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037332
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275355.1 NP_001262284.1 7 - 8 2358 68
NM_001300246.1 NP_001287175.1 7 - 8 2520 68
NM_001300247.1 NP_001287176.1 7 - 8 2376 68
NM_141285.2 NP_649542.1 7 - 8 2502 68
GenBank mRNA AY118365.1 - 7 - 8 2499 68

View Full Product Details