Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osi14
Entrez Gene ID
40770
Gene Name
Osiris 14
Gene Aliases
BEST:LD24139, CG1155, Dmel\CG1155, Dmel_CG1155, SP558, Sp558, osi14
Chromosome Location
Chr.3R: 6299128 - 6300738 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040279
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141378.3 NP_649635.1 1 - 2 364 76
GenBank mRNA AY058538.1 - 1 - 2 334 76

View Full Product Details