Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Desat2
- Entrez Gene ID
-
41536
- Gene Name
- Desaturase 2
- Gene Aliases
-
7-11HD, CG5925, Dmel\CG5925, Dmel_CG5925, Fad, desat, desat 2, desat-2, desat2, desat2/Fad, ds2
- Chromosome Location
-
Chr.3R: 12436248 - 12437857 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0043043
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141944.3 |
NP_650201.1 |
3 - 4 |
940 |
64 |
GenBank mRNA |
BT024427.1 |
- |
3 - 4 |
940 |
64 |
View Full Product Details