Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
yellow-f2
Entrez Gene ID
41596
Gene Name
CG8063-PA
Gene Aliases
CG8063, DI, Dmel\CG8063, Dmel_CG8063, yel-f2
Chromosome Location
Chr.3R: 12993395 - 12995147 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038105
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141990.3 NP_650247.1 2 - 3 598 62
GenBank mRNA AY060728.1 - 2 - 3 598 62

View Full Product Details