Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mf
Entrez Gene ID
41824
Gene Name
Myofilin
Gene Aliases
BcDNA:GH23305, CG6803, DmMf, Dmel\CG6803, Dmel_CG6803, Zeelin1, z1
Chromosome Location
Chr.3R: 15170625 - 15181510 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038294
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170138.1 NP_001163609.1 5 - 6 663 97
NM_001170140.1 NP_001163611.1 5 - 6 597 97
NM_001170141.1 NP_001163612.1 5 - 6 663 97
NM_001170142.1 NP_001163613.1 5 - 6 597 97
NM_001260184.1 NP_001247113.1 5 - 6 563 97
NM_001260185.1 NP_001247114.1 5 - 6 597 97
NM_001275656.1 NP_001262585.1 5 - 6 591 97
NM_001300405.1 NP_001287334.1 4 - 5 2413 97
NM_142166.2 NP_650423.1 5 - 6 597 97
NM_169613.1 NP_731970.1 5 - 6 597 97
GenBank mRNA AY058374.1 - 5 - 6 652 97
AY071198.1 - 5 - 6 597 97
AY113622.1 - 5 - 6 598 97
BT081982.1 - 5 - 6 654 97

View Full Product Details