Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mat89Ba
Entrez Gene ID
41973
Gene Name
Maternal transcript 89Ba
Gene Aliases
CG12785, Dmel\CG12785, Dmel_CG12785
Chromosome Location
Chr.3R: 16268329 - 16272415 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261286
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142273.4 NP_650530.2 3 - 4 1609 63
GenBank mRNA BT001511.1 - 3 - 4 1583 63
BT016113.1 - 3 - 4 1581 63

View Full Product Details