Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wrd
Entrez Gene ID
42169
Gene Name
well-rounded
Gene Aliases
B', B'/PR61, B56-1, BcDNA:GM05554, CG 7913, CG7901, CG7913, Dmel\CG7913, Dmel_CG7913, PP2A, PP2A B', PP2A-B, PP2A-B', PP2A[B'-1], Wrd, anon-WO0118547.420, dB56-1, dPP2A, dPP2A-B56-1, i234, pp2A-B'
Chromosome Location
Chr.3R: 18167757 - 18180049 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0042693
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144598.3 NP_001138070.1 2 - 3 544 84
NM_001144599.2 NP_001138071.1 2 - 3 544 84
NM_001260247.1 NP_001247176.1 2 - 3 676 84
NM_001260248.1 NP_001247177.1 2 - 3 503 84
NM_001260249.1 NP_001247178.1 2 - 3 950 84
NM_001260250.1 NP_001247179.1 1 - 2 1649 84
NM_142424.3 NP_650681.2 2 - 3 717 84
NM_169787.2 NP_732293.1 2 - 3 544 84
GenBank mRNA BT021232.1 - 2 - 3 543 84

View Full Product Details