Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mekk1
Entrez Gene ID
42253
Gene Name
CG7717-PB
Gene Aliases
CG7717, D-MEKK1, D-Mekk1, DmMEKK1, Dmel\CG7717, Dmel_CG7717, MEKK, MEKK1, MEKK4, Mekk1/4, dMEKK, dMEKK1, dMEKK4, dMekk1, mekk1, mekk1a, mekk1b
Chromosome Location
Chr.3R: 18733106 - 18743594 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024329
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142493.3 NP_650750.4 4 - 5 2295 64
NM_169833.2 NP_732373.1 4 - 5 2089 64
GenBank mRNA AY118659.1 - 4 - 5 2055 64

View Full Product Details