Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp12a4
- Entrez Gene ID
-
42294
- Gene Name
- CG6042-PA
- Gene Aliases
-
12a4, CG6042, CYP12A4, Cyp12A4, Dmel\CG6042, Dmel_CG6042, P450, Rst(Luf), cyp12a4
- Chromosome Location
-
Chr.3R: 19135158 - 19137136 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038681
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142526.4 |
NP_650783.2 |
3 - 4 |
1063 |
82 |
GenBank mRNA |
BT056257.1 |
- |
3 - 4 |
1061 |
82 |
View Full Product Details