Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppcs
Entrez Gene ID
42295
Gene Name
Phosphopantothenoylcysteine synthetase
Gene Aliases
BcDNA:GH03502, CG5629, Dmel\CG5629, Dmel_CG5629, GH03502, PPCS, anon-WO0118547.401, dPPCS
Chromosome Location
Chr.3R: 19139054 - 19147910 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261285
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316504.1 NP_001303433.1 3 - 4 1410 62
NM_142527.3 NP_650784.3 3 - 4 1410 62
NM_169848.3 NP_732401.1 3 - 4 1410 62
GenBank mRNA AF132150.1 - 3 - 4 1370 62

View Full Product Details