Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ir92a
Entrez Gene ID
42415
Gene Name
Ionotropic receptor 92a
Gene Aliases
CG15685, DmelIR92a, Dmel\CG15685, Dmel_CG15685, IR92a
Chromosome Location
Chr.3R: 20338973 - 20343902 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038789
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104375.2 NP_001097845.2 5 - 6 926 69

View Full Product Details