Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Npc2f
- Entrez Gene ID
-
42864
- Gene Name
- Niemann-Pick type C-2f
- Gene Aliases
-
BcDNA:LP08842, CG6164, Dmel\CG6164, Dmel_CG6164, npc2f
- Chromosome Location
-
Chr.3R: 24137895 - 24140936 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039154
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142962.3 |
NP_651219.1 |
2 - 3 |
417 |
63 |
GenBank mRNA |
AY061552.1 |
- |
2 - 3 |
417 |
63 |
View Full Product Details