Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nct
Entrez Gene ID
42964
Gene Name
Nicastrin
Gene Aliases
CG7012, Dmel\CG7012, Dmel_CG7012, Nic, S5A, ST96B, agro, dNct, nct, nic
Chromosome Location
Chr.3R: 24851601 - 24854869 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039234
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170242.2 NP_001163713.1 5 - 6 1913 84
NM_001276003.1 NP_001262932.1 5 - 6 1913 84
NM_143040.5 NP_651297.2 5 - 6 1913 84
NM_206568.4 NP_996291.2 5 - 6 1913 84

View Full Product Details