Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clbn
Entrez Gene ID
43018
Gene Name
Caliban
Gene Aliases
CG11847, Dmel\CG11847, Dmel_CG11847, clbn
Chromosome Location
Chr.3R: 25127502 - 25131155 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259152
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170250.1 NP_001163721.1 1 - 2 242 76
NM_143084.2 NP_651341.2 2 - 3 230 76
GenBank mRNA BT030842.1 - 1 - 2 180 76

View Full Product Details