Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHKov2
Entrez Gene ID
43068
Gene Name
CHK domain ov2
Gene Aliases
CG10675, Dmel\CG10675, Dmel_CG10675, ref(3)D
Chromosome Location
Chr.3R: 25329530 - 25331203 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039328
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143130.4 NP_651387.1 4 - 5 1100 69
GenBank mRNA AY058384.1 - 4 - 5 1079 69

View Full Product Details